missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Honeywell™ Fluka™

£41.12 - £480.31

Specifications

UN Number UN1230
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15664420
View Documents
Honeywell-Hydranal
34801-500ML
500 mL
£41.12
500mL
Please to purchase this item. Need a web account? Register with us today!
15634420
View Documents Promotion Details
Honeywell-Hydranal
34801-1L
1 L
£57.47
1 L
Please to purchase this item. Need a web account? Register with us today!
15644420
View Documents
Honeywell-Hydranal
34801-2.5L
2.5 L
£137.79
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15684420
View Documents
Honeywell Fluka
34801-6X500ML
6 x 500 mL
£214.85
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15674420
View Documents
Honeywell Fluka
34801-6X1L
6 x 1 L
£300.07
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15654420
View Documents
Honeywell Fluka
34801-4X2.5L
4 x 2.5 L
£480.31
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

UN1230
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310 + P330-P308 + P311-P403 + P233

Product Identifier
  • HYDRANAL Titrant 5

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol, iodine

SDS
Documents

Documents

Product Certifications