Learn More
3M™ Hydrated-Sponge
Uses simplified packaging to minimize handling and preparation, and reduces waste.
Brand: 3M™ 7100067517
Additional Details : CAS Number : 7732-18-5 Weight : 0.19800kg
Description
- Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection.
- Durable sponge withstands scrubbing.
- Sponge bag with write-on label for ease in labeling samples.
- Includes two decontaminated gloves for safety and convenience.
Specifications
Hydrated Sponge | |
7.6 cm | |
4 cm | |
10 mL Letheen Broth HS10LET |
3 in. | |
1.5 in. | |
100/Cs. |