missing translation for 'onlineSavingsMsg'
Learn More
Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals Product Code.: 10521794

Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals

Product Code. 10521794
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10521794

Brand: Thermo Scientific Acros 296811000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10217-52-4, 7732-18-5
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739Show MoreShow Less
PubChem CID 24654
ChEBI CHEBI:35511
SMILES NN

Specifications

Boiling Point 109.4°C
Melting Point -65.0°C
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 55%
Linear Formula H2NNH2·xH2O
Fieser 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174
Flash Point >100°C
Merck Index 14,4771
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether
IUPAC Name hydrazine;hydrate
Formula Weight 32.04
Percent Purity 55% (Hydrazine,35%)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3910
Specific Gravity 1.023
Density 1.0230g/mL
Chemical Name or Material Hydrazine hydrate, 55% (Hydrazine 35%)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457836 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number296810025,296815000,296811000
Lot NumberA0356757 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number296810025,296815000,296811000
Lot NumberA0415895 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number296810025,296815000,296811000
Lot NumberA0440567 Certificate TypeCertificate of Analysis Date30/03/2022 Catalog Number296810025,296815000,296811000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine hydrate, 55% (Hydrazine, 35%)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Hydrazine (hydrate),Hydrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrazine hydrate, 55% (Hydrazine, 35%), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.