missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent

Specifications
Specifications
Melting Point | 89°C to 93°C |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 98% min. (Argentometry) |
Linear Formula | H2NNH2·HCl |
Solubility Information | Solubility in water: 370g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in most organic solvents,soluble in liquid ammoniak |
Formula Weight | 68.51 |
Percent Purity | ≥98% |
Infrared Spectrum | Authentic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465174 | Certificate of Analysis | 15/08/2024 | 201992500,201990010,201990051 |
A0459399 | Certificate of Analysis | 11/06/2024 | 201992500,201990010,201990051 |
A0452036 | Certificate of Analysis | 02/05/2023 | 201992500,201990010,201990051 |
A0400372 | Certificate of Analysis | 14/12/2022 | 201992500,201990010,201990051 |
A0418604 | Certificate of Analysis | 12/12/2022 | 201992500,201990010,201990051 |
Safety and Handling



Product Identifier
- Hydrazine monohydrochloride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350-May cause cancer.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction