missing translation for 'onlineSavingsMsg'
Learn More
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals Product Code.: 10578902

Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals

Product Code. 10578902
100 g, Plastic bottle
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10578902

Brand: Thermo Scientific Acros 423771000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H4N2·H2SO4
Molecular Weight (g/mol) 130.12
MDL Number MFCD00044873
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
Synonym hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfateShow MoreShow Less
PubChem CID 24842
IUPAC Name hydrazine;sulfuric acid
SMILES NN.OS(=O)(=O)O

Specifications

Melting Point 254.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula H2NNH2·H2SO4
Merck Index 15, 4810
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 130.12
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.05% max
Chemical Name or Material Hydrazine sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467251 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number423771000,423775000,423770500
Lot NumberA0467252 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number423771000,423775000,423770500
Lot NumberA0448765 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number423771000,423775000,423770500
Lot NumberA0463597 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number423771000,423775000,423770500
Lot NumberA0456350 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number423771000,423775000,423770500
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrazine sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.