missing translation for 'onlineSavingsMsg'
Learn More
Hydrazinium Chloride, 98+%, Extra Pure, SLR, Fisher Chemical Product Code.: 10530151

Hydrazinium Chloride, 98+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10530151
250 g, Plastic powder jar
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10530151

Brand: Fisher Chemical H/0600/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 200°C
Melting Point 198°C
CAS 5341-61-7
Color White
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula ClH5N2
MDL Number MFCD00064543
Synonym hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
InChI Key BIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES [H+].[Cl-].NN
IUPAC Name hydrazine;dihydrochloride
Molecular Weight (g/mol) 68.50
PubChem CID 17548
Formula Weight 104.97g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrazinium Chloride, 98+%, Extra Pure, SLR, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.