missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydriodic Acid, no stabilizer, Honeywell
£41.29 - £133.92
Chemical Identifiers
CAS | 10034-85-2 |
---|---|
Molecular Formula | HI |
Molecular Weight (g/mol) | 127.91 |
MDL Number | MFCD00011347 |
InChI Key | XMBWDFGMSWQBCA-UHFFFAOYSA-N |
Synonym | hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group |
PubChem CID | 24841 |
ChEBI | CHEBI:43451 |
IUPAC Name | hydrogen iodide |
SMILES | I |
Description
Chemical Identifiers
10034-85-2 | |
127.91 | |
XMBWDFGMSWQBCA-UHFFFAOYSA-N | |
24841 | |
hydrogen iodide |
HI | |
MFCD00011347 | |
hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group | |
CHEBI:43451 | |
I |
Specifications
10034-85-2 | |
HI | |
1787 | |
XMBWDFGMSWQBCA-UHFFFAOYSA-N | |
hydrogen iodide | |
24841 | |
127.91g/mol | |
Hydriodic Acid, Distilled, Contains No Stabilizer |
1.701g/mL at 25°C | |
MFCD00011347 | |
hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group | |
I | |
127.91 | |
CHEBI:43451 | |
99.99% (trace metals basis) |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314-H335
Danger
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title