missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 0.01N Standardized Solution, Thermo Scientific Chemicals
£23.90 - £114.00
Chemical Identifiers
CAS | 7647-01-0, 7732-18-5 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303797
|
Thermo Scientific Alfa Aesar
035648.K2 |
1 L |
£23.90
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393787
|
Thermo Scientific Alfa Aesar
035648.K7 |
4 L | |||||||
11383787
|
Thermo Scientific Alfa Aesar
035648.M4 |
20 L |
£114.00
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrochloric acid, 0.01N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-01-0, 7732-18-5 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
CHEBI:17883 |
ClH | |
MFCD00011324 MFCD00792839 | |
313 | |
Cl |
Specifications
7647-01-0, 7732-18-5 | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
Colorless | |
MFCD00011324 MFCD00792839 | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid: 0.083%; Water: 99.917% | |
Miscible with water, methanol,ethanol and ether | |
Hydrochloric acid, 0.01N |
Safety and Handling
missing translation for 'einecsNumber' : 231-595-7
missing translation for 'rtecsNumber' : MW4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only