missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
6 x 500 mL
4 x 4 L
189 L
Density:
1.2 g/mL at 25°C
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Concentration | 37% |
Density | 1.2 g/mL at 25°C |
Residue after Ignition | ≤5ppm |
Vapor Pressure | 3.23psi (21.1°C), 7.93psi (37.7°C) |
Quantity | 4 x 4 L |
UN Number | 1789 |
Beilstein | 1098214 |
IUPAC Name | chlorane |
Formula Weight | 36.46g/mol |
Grade | ACS Reagent |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Safety and Handling


Product Identifier
- Hydrochloric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction