missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, ACS Reagent, 37%, Honeywell Fluka™
ACS Reagent, 37%
£19.72 - £1007.64
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Density | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Density | Price | Quantity & Availability | ||||
15638350
|
Honeywell-Fluka
258148-25ML |
25 mL | 1.2 g/mL at 25°C | N/A | |||||
15602790
|
Honeywell-Fluka
258148-100ML |
100 mL | 1.2 g/mL at 25°C | N/A | |||||
15642790
|
Honeywell-Fluka
258148-500ML |
500 mL | 1.2 g/mL at 25°C |
£49.35
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15622790
|
Honeywell-Fluka
258148-2.5L |
2.5 L | 1.2 g/mL at 25°C |
£86.93
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15652790
|
Honeywell-Fluka
258148-6X500ML |
6 x 500 mL | 1.2 g/mL at 25°C |
£258.16
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15662790
|
Honeywell-Fluka
258148-6X500ML-GL |
6 x 500 mL | 1.2 g/mL at 25°C | N/A | |||||
15648350
|
Honeywell-Fluka
258148-4L |
4 L | 1.2 g/mL at 25°C |
£127.42
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15632790
|
Honeywell-Fluka
258148-4X4L |
4 x 4 L | 1.2 g/mL at 25°C |
£441.47
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15612790
|
Honeywell-Fluka
258148-189L |
189 L | 1.2 g/mL at 25°C | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
7647-01-0 | |
≤5ppm | |
ClH | |
1789 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
ACS Reagent |
37% | |
3.23psi (21.1°C), 7.93psi (37.7°C) | |
MFCD00011324 MFCD00792839 | |
1098214 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46g/mol | |
Hydrochloric Acid |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314-H335
missing translation for 'einecsNumber' : 231-595-7
missing translation for 'rtecsNumber' : MW4025000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrochloric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.