missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%, Honeywell Fluka™
Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥40%
£99.30 - £595.69
Chemical Identifiers
CAS | 7664-39-3 |
---|---|
Molecular Formula | FH |
Molecular Weight (g/mol) | 20.01 |
MDL Number | MFCD00011346 |
InChI Key | KRHYYFGTRYWZRS-UHFFFAOYSA-N |
Synonym | hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 |
PubChem CID | 14917 |
ChEBI | CHEBI:29228 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15602980
|
Honeywell-Fluka
30103-1L |
1 L |
£99.30
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15612980
|
Honeywell-Fluka
30103-2.5L |
2.5 L |
£195.54
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15642980
|
Honeywell-Fluka
30103-6X1L |
6 x 1 L |
£470.48
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15622980
|
Honeywell-Fluka
30103-4X2.5L |
4 x 2.5 L |
£595.69
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7664-39-3 | |
20.01 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
14917 |
FH | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
CHEBI:29228 |
Specifications
7664-39-3 | |
1.15g/mL at 25°C | |
25mmHg (20°C) | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
F | |
20.01 | |
CHEBI:29228 | |
ACS/puriss. p.a. |
≥40% | |
≤0.0005% (as SO4) | |
FH | |
1790 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
hydrogen fluoride | |
14917 | |
20.01g/mol | |
Hydrofluoric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
H300 + H310 + H330-H314
missing translation for 'einecsNumber' : 231-634-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrofluoric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen fluoride