missing translation for 'onlineSavingsMsg'
Learn More
Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals Product Code.: 10065760

Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals

Product Code. 10065760
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10065760

Brand: Thermo Scientific Acros 411885000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Undesignated
Boiling Point 108.0°C
Melting Point -33.0°C
Linear Formula HOOH
Fieser 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Density 1.1100g/mL
Merck Index 15, 4835
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ether, alcohol, insoluble in petroleum ether, decomposed by many organic solvents
Formula Weight 34
Percent Purity 29.0 to 32.0%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Evaporation 0.002% max.
Specific Gravity 1.11
Decomposition Information >125°C
Quantity 500 g
Physical Form Solution
Chemical Name or Material Hydrogen peroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451292 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number411885000,411880025,411881000,411880010
Lot NumberA0445712 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number411885000,411880025,411881000,411880010
Lot NumberB0547445A Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number411885000,411880025,411881000,411880010
Lot NumberA0445712 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog Number411885000,411880025,411881000,411880010
Lot NumberA0451292 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number411885000,411880025,411881000,411880010
compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide 20-35wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    29

    Hours

    1

    Minutes

    29

    Seconds

    58

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.