Learn More
Hydrogen peroxide, pure, 50 wt% solution in water, stabilized, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Boiling Point | 114.0°C |
Melting Point | -52.0°C |
Linear Formula | H2O2 |
Fieser | 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145 |
Density | 1.2000g/mL |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether |
Formula Weight | 34 |
Percent Purity | 49.8 to 50.6% |
Grade | Pure |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467360 | Certificate of Analysis | 08/11/2024 | 302860025,302865000 |
A0460379 | Certificate of Analysis | 19/02/2024 | 302860025,302865000 |
A0454790 | Certificate of Analysis | 25/01/2024 | 302860025,302865000 |
A0448055 | Certificate of Analysis | 25/08/2023 | 302860025,302865000 |
A0395294 | Certificate of Analysis | 14/12/2022 | 302860025,302865000 |
Safety and Handling



- Hydrogen peroxide, 50 wt% solution in water
- Danger
- Oxidising liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrogen peroxide
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
27Hours
7Minutes
10Seconds
20 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
58Hours
7Minutes
10Seconds
20New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.