Learn More
Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals
£10.90 - £198.99
Chemical Identifiers
CAS | 10039-54-0 |
---|---|
Molecular Formula | H6N2O2·H2SO4 |
MDL Number | MFCD00044869 |
Synonym | Hydroxylammonium sulfate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10286920
|
Thermo Scientific Acros
198530050 |
5 g | Glass Bottle | N/A | ||||||
10701861
|
Thermo Scientific Acros
198531000 |
100 g | Plastic bottle |
£20.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10722051
|
Thermo Scientific Acros
198530010 |
1 kg | Plastic bottle |
£49.65
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10464025
|
Thermo Scientific Acros
198530051 |
5 kg | Plastic bottle |
£198.99
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10039-54-0 | |
H6N2O2·H2SO4 | |
MFCD00044869 | |
Hydroxylammonium sulfate |
Specifications
10039-54-0 | |
Crystals | |
H6N2O2·H2SO4 | |
MFCD00044869 | |
15, 4867 | |
164.14 | |
Authentic |
White | |
99% | |
(H2NOH)2·H2SO4 | |
01,481 | |
Hydroxylammonium sulfate | |
≥98.5% | |
Hydroxylamine sulfate |
Safety and Handling
GHS P Statement
May be corrosive to metals.
Suspected of causing cancer if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.