missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Hydroxypropyl cellulose, Average M.W. 100.000 Product Code.: 10477380

Thermo Scientific Chemicals Hydroxypropyl cellulose, Average M.W. 100.000

Product Code. 10477380
250 g, Glass bottle
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10477380

Brand: Thermo Scientific Chemicals 184882500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
MDL Number MFCD00132688
Synonym 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?cShow MoreShow Less

Specifications

Color White to Cream
pH 5 to 8.5 (1% soln.)
Physical Form Granules or Crystalline Powder
Quantity 250 g
Formula Weight 100.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 5% max. (105°C)
Packaging Glass bottle
Viscosity 75-150 mPa.s 5% aq.sol.
Chemical Name or Material Hydroxypropyl cellulose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462573 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number184880051,184880010,184881000,184882500
Lot NumberA0425016 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number184880051,184880010,184881000,184882500
Lot NumberA0400396 Certificate TypeCertificate of Analysis Date22/03/2021 Catalog Number184880051,184880010,184881000,184882500
Product Identifier
  • Hydroxypropyl cellulose, Average M.W. 100.000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Hydroxypropyl cellulose, Average M.W. 100.000 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.