Learn More
Ansell HyFlex™ 11-132 Anti-Static Lint-Free Polyamide Gloves
Gloves are knitted using a seamless non-stop process with a textile liner made of copper-doped polyamide thread. Anti-static properties protect users from electrostatic discharge.
£27.15
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Seamless Knit
Color Natural
Hand Ambidextrous
Material Intercept Technology Yarn
View More Specs
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16009591
View Documents Promotion Details
Ansell
11132060
6 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16019591
View Documents Promotion Details
Ansell
11132070
7 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16029591
View Documents Promotion Details
Ansell
11132080
8 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16039591
View Documents Promotion Details
Ansell
11132090
9 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16049591
View Documents Promotion Details
Ansell
11132100
10
£27.15
12 pairs
In Stock
to see stock.
Add to basket
Description
Description
  • Knitted with a seamless, non-stop process
  • Elastic polyamide textile liner doped with copper
  • Antistatic properties protect the user against electrostatic discharge
  • Ambidextrous shape doubles the life of the glove by enabling the use of both sides
  • Lint free gloves feature fine, stretchable stitches
Specifications
Specifications
EN 420:2003 + A1:2009
Natural
Intercept Technology Yarn
Knit
Seamless Knit
Ambidextrous
1131A
11-132