missing translation for 'onlineSavingsMsg'
Learn More
Hygromycin B from Streptomyces Hygroscopicus, MP Biomedicals™ Product Code.: 11470722
compliance-icons compliance-icons compliance-icons

Hygromycin B from Streptomyces Hygroscopicus, MP Biomedicals™

Product Code. 11470722 Shop All MP Biomedicals Products
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 11470722

Brand: MP Biomedicals 0219417091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hygromycin B is an antibiotic substance isolated from Streptomyces hygroscopicus.

Its mode of action is the inhibition of protein synthesis, by inducing the misreading of the m-RNA template in the prokaryote, (E. coli, at 100 mg/mL), lower eukaryotes (e.g., yeast, at 200mg/mL) and higher eukaryotes (e.g., mammalian cells in culture). It selectively penetrates cells that have been rendered permeable by virus infection and combined with its potency to inhibit translation, it is an effective antiviral agent. Hygromycin B can be used as a dual-selection antibiotic when used in conjunction with G418 or Zeocin™ for cells transfected with two different vectors.

  • Blocks polypeptide synthesis and inhibits elongation. Specifically, it inhibits protein synthesis by interfering with translocation of the 70S ribosome and inducing misreading of the mRNA template.
  • When using this product for mammalian cell selection the working concentration is generally between 50mg/mL and 1mg/mL, but at times may go as high as 1 mg/mL.
  • The appropriate concentration for use must be determined empirically for each cell type.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Powder
CAS 31282-04-9
Melting Point 160°C to 180°C (literature)
Quantity 1 g
Molecular Formula C20H37N3O13
Synonym O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene- (1->2-3)-O-b-D- talopyranosyl- (1->5)- 2-deoxy- N3-methyl-D-streptamine
Solubility Information Freely soluble in water (50 mg/mL), methanol,buffer solution and ethanol. Practically insoluble in less polar solvents.
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
Molecular Weight (g/mol) 527.524
PubChem CID 134129613
Formula Weight 527.52g/mol
Percent Purity ∼95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hygromycin B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H300 + H330-Fatal if swallowed or if inhaled.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hygromycin B from Streptomyces Hygroscopicus, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.