Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.
Brand: Thermo Scientific™ SH30647.19
Additional Details : Weight : 0.45000kg
Description
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
28 L | |
Cardboard Cartons | |
Irradiated | |
CX3-9 |
7.5 gal. | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
HyPerforma |