missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Polystyrene Lab Plates

Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.

Brand:  Thermo Scientific™ 60110-207

 View more versions of this product

Product Code. 10138074

This item is not returnable. View return policy

Description

Description

Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:

  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography

The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Titanium Dioxide; Polystyrene
HyperSep
96-Well Plate
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep Titanium Dioxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers