Learn More
Thermo Scientific™ HyperSep™ Polystyrene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
Brand: Thermo Scientific™
Code : 16
Additional Details : Weight : 0.50000kg
Description
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:
- Tissue culture and separation of products
- Concentration of small-scale samples
- Sample clean-up
- Collection of sample after chromatography
Specifications
Zirconium Dioxide; Polystyrene | |
HyperSep | |
96-Well Plate |
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules | |
HyperSep Zirconium Dioxide |