missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Retain PEP Plates

Get balanced retention for polar and nonpolar analytes with HyperSep Retain PEP Plates.

Brand:  Thermo Scientific™ 60306-208

 View more versions of this product

Product Code. 13416976

  • £364.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Retain PEP 60 mg/2 mL 96 Fixed Well Plate, 1 pk
Retention of Polar and Nonpolar Analytes
HyperSep
Solid Phase Extraction Well Plate
1 Pack
60 mg
30 to 50 μm
HyperSep Urea-Modified Polystyrene DVB
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.