missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HyperSep™ Retain PEP Plates
Get balanced retention for polar and nonpolar analytes with HyperSep Retain PEP Plates.
£294.23 - £352.77
Specifications
For Use With (Application) | Retention of Polar and Nonpolar Analytes |
---|---|
Particle Size | 30 to 50 μm |
Product Line | HyperSep |
Sorbent | HyperSep Urea-Modified Polystyrene DVB |
Type | Solid Phase Extraction Well Plate |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11507671
|
Thermo Scientific™
60306-207 |
Retain PEP 30 mg/2 mL 96 Fixed Well Plate, 1 pk |
£352.77
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
13416976
|
Thermo Scientific™
60306-208 |
Retain PEP 60 mg/2 mL 96 Fixed Well Plate, 1 pk |
£342.72
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
10258124
|
Thermo Scientific™
60303-207 |
Retain PEP 30 mg/1 mL 96 Removable Well Plate, 1 pk |
£294.23
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
Description
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Plates, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.- [bullet] High, consistent recoveries
- [bullet] Fast and easy sample preparation and method development
- [bullet] Reproducible, reliable sample preparation in a traditional format
- [bullet] Available in a wide range of chemistries and bed weights
- [bullet] Drugs and metabolites in biological fluids
- [bullet] Peptides in serum, plasma or biological fluids
- [bullet] Environmental samples
Specifications
Retention of Polar and Nonpolar Analytes | |
HyperSep | |
Solid Phase Extraction Well Plate | |
1 Pack |
30 to 50 μm | |
HyperSep Urea-Modified Polystyrene DVB | |
1 mL |