Learn More
Thermo Scientific™ HyperSep™ Silica Cartridges
Retain analytes in nonpolar matrices, and separate compounds of very similar structure with these silica SPE cartridges.
£107.54 - £753.75
Specifications
For Use With (Application) | Retention of Analytes in Nonpolar Matrices |
---|---|
Particle Size | 40 to 60 μm |
Product Line | HyperSep |
Type | Solid Phase Extraction Cartridge |
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Sorbent | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Sorbent | Quantity | Price | Quantity & Availability | ||||||
10553985
|
Thermo Scientific™
60108-315 |
500 mg | 3 mL | HyperSep Silica | Pack of 50 |
£184.00
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10563985
|
Thermo Scientific™
60108-317 |
100 mg | 1 mL | HyperSep Silica | Pack of 100 |
£187.00
Pack of 100 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10036414
|
Thermo Scientific™
60108-409 |
50 mg | 1 mL | HyperSep Silica | Pack of 100 |
£210.00
Pack of 100 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
10672284
|
Thermo Scientific™
60108-410 |
200 mg | 3 mL | HyperSep Silica | Pack of 50 |
£107.54
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10120743
|
Thermo Scientific™
60108-411 |
500 mg | 6 mL | HyperSep Silica | Pack of 30 |
£120.64
Pack of 30 |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10414603
|
Thermo Scientific™
60108-426 |
1000 mg | 6 mL | HyperSep Silica | Pack of 30 |
£136.00
Pack of 30 |
In Stock
Log in to see stock. |
||||||
10075674
|
Thermo Scientific™
60108-710 |
2000 mg | 15 mL | HyperSep Silica | Pack of 20 |
£139.70
Pack of 20 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
10436934
|
Thermo Scientific™
60108-711 |
5000 mg | 25 mL | HyperSep Silica | Pack of 20 |
£129.65
Pack of 20 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
10514105
|
Thermo Scientific™
60108-712 |
10 g | 75 mL | HyperSep Silica | Pack of 10 |
£166.00
Pack of 10 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
15373426
|
Thermo Scientific™
60108-503 |
100 mg | 10 mL | HyperSep Silica | Pack of 50 |
£177.89
Pack of 50 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
13496936
|
Thermo Scientific™
60108-852 |
70,000 mg | 150 mL | HyperSep Silica | Pack of 10 | N/A | N/A | N/A | |||||
13382120
|
Thermo Scientific™
60108-793 |
500 mg | 10 mL | HyperSep Silica | Pack of 50 |
£156.78
Pack of 50 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
13456826
|
Thermo Scientific™
60108-851 |
20,000 mg | 75 mL | HyperSep Silica | Pack of 10 |
£232.16
Pack of 10 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
13406946
|
Thermo Scientific™
60108-850 |
50,000 mg | 150 mL | HyperSep Silica | Pack of 10 | N/A | N/A | N/A | |||||
13416946
|
Thermo Scientific™
60108-853 |
10 g | 25 mL | HyperSep Silica | Pack of 20 |
£234.17
Pack of 20 |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
Beschreibung
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ Silica Cartridges feature a polar sorbent primarily used to retain analytes in nonpolar matrices, and can effectively separate compounds of very similar structure. Typical applications include:
- Extraction of polar compounds including aldehydes, amines, drugs, pesticides and herbicides
- Extraction of carotenoids, fat-soluble vitamins, aflatoxins in food matrices
- Extraction of fatty acids and phospholipids
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Spezifikation
Retention of Analytes in Nonpolar Matrices | |
40 to 60 μm | |
HyperSep | |
Solid Phase Extraction Cartridge |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.