Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
Brand:  Thermo Scientific™ 60109-406
 View more versions of this product
Product Code. 10304673
Description
Description
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • [bullet] Filter-free design to eliminate dead volume
  • [bullet] No contamination from the supporting matrix
  • [bullet] Available in many popular chemistries
Applications include:
  • [bullet] Mass spectrometry
  • [bullet] Desalting
  • [bullet] Protein purification
  • [bullet] MALDI
  • [bullet] Electrophoresis
  • [bullet] HPCE
  • [bullet] HPLC
  • [bullet] CEC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HILIC; 1-10 uL Vol.
Micropipettes
HyperSep HILIC
1 to 10 μL
Microscale Solid Phase Extraction
HyperSep
SpinTip
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 1-10μL >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.