missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hypersil™ MOS C8 LC Column, 5μm
Reliably analyze moderately hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids and environmental samples.
Brand: Thermo Scientific™ 30205-124630
Code : 16
Additional Details : Weight : 0.10000kg
Description
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Specifications
Analytical Column | |
125 mm | |
5 μm | |
120 Å | |
60°C | |
C8 | |
6.5% | |
No |
4.6 mm | |
5800 psi (400 bar) | |
2 to 8 | |
170 m²/g | |
L7 | |
Spherical, Fully Porous Type A Silica | |
Reversed Phase | |
Reversed Phase |