missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hypersil™ MOS C8 LC Column, 5μm
Reliably analyze moderately hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids and environmental samples.
£412.08 - £629.27
Specifications
Column Format | Analytical Column |
---|---|
Particle Size | 5 μm |
pH | 2 to 8 |
Pore Size | 120 Å |
Surface Area | 170 m²/g |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11597461
|
Thermo Scientific™
30205-204630 |
4.6 mm | 200 mm |
£602.17
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11587461
|
Thermo Scientific™
30205-202130 |
2.1 mm | 200 mm |
£629.27
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11309081
|
Thermo Scientific™
30205-124630 |
4.6 mm | 125 mm |
£571.05
Each |
In Stock
Log in to see stock. |
||||
10334423
|
Thermo Scientific™
30205-102130 |
2.1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Specifications
Analytical Column | |
2 to 8 | |
170 m²/g | |
C8 | |
Reversed Phase |
5 μm | |
120 Å | |
L7 | |
6.5% | |
Reversed Phase |