Learn More
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Make procedures as painless as possible with extremely sharp and reliable stainless steel needles. Terumo™ Agani™ Single-use Sterile Hypodermic Needles feature a unique, latex-free double bevel design for maximum comfort.
Brand:  Terumo™ 8AN2716R1
 View more versions of this product
Product Code. 15331557
  • £7.00 / Pack of 100
Edge
Explore more special offers
Description
Description
  • Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
  • All needles are supplied sterile wrapped in individual, tamper proof peel pouches
  • Simple color coded system allows easy identification of required sizes
  • Use with a wide range of Sterile Syringes
  • Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
  • CE marked and latex free
  • Supplied in boxes of 100 needles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Class IIa, sterile product following MDD 93/42/EEC
Gray
0.4 mm
16 mm
Single-use
11° Regular
Sterile
Dimethyl Silicone Oil, Epoxy Resin, Polypropylene, Stainless Steel
27 G
Yes
Sterile Hypodermic Needle for Single-Use
Box of 100
Sterile Syringes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Terumo™ Agani™ Single-use Sterile Hypodermic Needles > Grey; Gauge: 27G; Length: 16mm; Needle Point Style: Regular, 11°