missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Clear Autoclave Bags

Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.

Brand:  Fisherbrand™ FB60428

Product Code. 11563342

This item is not returnable. View return policy

Description

Description

  • For high temperature sterilisation (134°C) to a maximum of 140°C
  • 40μm thickness
  • High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
  • For safe dissposal of biohazard waste
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Printed with a biohazard warning
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Polypropylene Clear Autoclave Bags > 620W x 780mmD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.