missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyQ™-RS Reduced Serum Media
For applications requiring a lower concentration of serum
Brand: Cytiva SH30564.01
24.08 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Support cell culture growth
- Format: Liquid
- Store at 2° to 8°C
Specifications
2° to 8°C | |
Liquid | |
0.1 μm Sterile-filtered | |
500 mL |
MEM-RS with L-glutamine | |
Reduced Serum Medium | |
Glutamine, Glucose |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction