Learn More
BUCHI I-180 Controller for Rotavapor R-80
The I-180 is an easy-to-use interface for conveniently regulating your vacuum automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
Marke: BUCHI 11I1800
Beschreibung
- Functional design fulfills essential needs of vacuum regulation
- Saves resources thanks to control of vacuum pump
- High longevity due to robust and durable design
- User-friendly and intuitive handling due to self-explaining user interface
- Easy and accurate vacuum regulation
- Relevant information is displayed at a glance
- High reliability due to centralized control of major process parameters
- Full process control thanks to parameter monitoring
- Proven technology for major applications
Spezifikation
Controller Interface | |
Rotavapor R-80 V-80/180 House Vacuum | |
Valve Unit, Woulff Bottle, Communication Cable, Tubing |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.