Learn More
Tocris Bioscience™ I-BET 762
Potent and high affinity BET bromodomain inhibitor; anti-inflammatory; orally bioavailable.
Brand:  Tocris Bioscience™ 6521/5
74.37 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18054232
  • £94.00 / 5mg
Explore more special offers
Description
Description
I-BET 762 is a potent and high affinity BET bromodomain inhibitor (IC50 = 32.5 - 42.5 nM; Kd = 50.5 - 61.3 nM). Inhibits myeloma cell proliferation. Attenuates transcription of oncogenic MYC and suppresses key inflammatory genes. Exhibits efficacy in a range of oncology and immunoinflammatory models. Displays antiproliferative effect in vitro and in vivo. Orally bioavailable.
Specifications
Specifications
(4S)-6-(4-Chlorophenyl)-N-ethyl-8-methoxy-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-4-acetamide
5 mg
C22H22ClN5O2
1260907-17-2
Bromodomain Inhibitors
0.98