missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ I-BET 762
Potent and high affinity BET bromodomain inhibitor; anti-inflammatory; orally bioavailable.
£94.00 - £367.00
Specifications
Chemical Name or Material | (4S)-6-(4-Chlorophenyl)-N-ethyl-8-methoxy-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-4-acetamide |
---|---|
CAS | 1260907-17-2 |
Target | Bromodomain Inhibitors |
Molecular Formula | C22H22ClN5O2 |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18094322
|
Tocris Bioscience™
6521/25 |
25 mg |
£367.00
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18054232
|
Tocris Bioscience™
6521/5 |
5 mg |
£94.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
I-BET 762 is a potent and high affinity BET bromodomain inhibitor (IC50 = 32.5 - 42.5 nM; Kd = 50.5 - 61.3 nM). Inhibits myeloma cell proliferation. Attenuates transcription of oncogenic MYC and suppresses key inflammatory genes. Exhibits efficacy in a range of oncology and immunoinflammatory models. Displays antiproliferative effect in vitro and in vivo. Orally bioavailable.Specifications
(4S)-6-(4-Chlorophenyl)-N-ethyl-8-methoxy-1-methyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-4-acetamide | |
Bromodomain Inhibitors | |
0.98 |
1260907-17-2 | |
C22H22ClN5O2 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title