Learn More
Ambion™ mirVana™ miRNA Mimic
Small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity.
Brand:  Ambion™ 4464075
24952.80 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
 View more versions of this product
Product Code. 13493079
  • £33720.00 / Each
Explore more special offers
Description
Description
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. mirVana™ miRNA Mimics are more specific than their predecessors due to proprietary chemical modifications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Room temperature
miRNA
1 nmol
mirVana miRNA mimics Library, 1nmol, Small, Chemically Modified Double-stranded RNAs that Mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-regulation of miRNA Activity
mirVana™ Mimics and Inhibitors
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ambion™ mirVana™ miRNA Mimic > 1 nmol