missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ I/P™ Standard Pump Head for I/P™ Tubing
Best accuracy and repeatability of any I/P pump head—ideal for dispensing and metering
Brand: Masterflex™ 07019-53
This item is not returnable.
View return policy
Description
- Each model is optimized for a single tube size to maximize performance and tubing life
- The cost-effective pump head is an excellent choice for dedicated applications
- Stack two pump heads to double the flow rate
- The Standard pump head’s classic design ensures the most accurate and repeatable performance—important in dispensing applications.
- A 360-degree precision-molded occlusion body ensures optimum accuracy and long tubing life.
- The design of the standard pump head enables the most precise dispensing accuracy of any MASTERFLEX pump head
Specifications
I/P Standard pump head for I/P 82 tubing, PPS housing/SS rotor. | |
Polyphenylene Sulfide | |
3 | |
5.5 in. | |
3.312 in. | |
5.2 in. | |
Stainless Steel | |
Volume Per Revolution With I/P 82 Tubing (mL): 20mL | |
0.1 to 650 rpm |
13 L/min. | |
1 | |
Fixed | |
139 mm | |
85 mm | |
132 mm | |
82 | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction