Learn More
Thermo Scientific™ I-PER™ Insect Cell Protein Extraction Reagent
Extract soluble protein from baculovirus-infected insect cells grown in suspension or monolayer culture with this gentle and effective lysis reagent.
Brand: Thermo Scientific™ 89802
256.50 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ I-PER Insect Cell Protein Extraction Reagent enables gentle and effective extraction of soluble protein from baculovirus-infected insect cells grown in suspension or monolayer culture.
The baculovirus insect cell expression system is an efficient and popular system for the production of recombinant (eukaryotic) proteins in cell culture. Proteins expressed in baculoviral systems can be used for structural analyses, biochemical assays and a variety of other applications. I-PER Reagent maintains functionality of extracted proteins and is directly compatible with downstream applications such as protein assays, Western blotting and 6xHis-tagged protein purification.Highlights:
Gentle extraction – optimized, mild non-ionic detergent provides maximum extraction of soluble proteins from insect cells
Effective – provides better protein extraction than sonication
Compatible – downstream compatibility with Western blotting, 6xHis-tagged protein purification, protein assays and ion-exchange chromatography
Flexible – useful for protein extraction from suspended or adherent cultured insect cells

Specifications
250 mL | |
Reagent | |
Upon receipt store at 4°C. |
Liquid | |
250 mL | |
I-PER™ |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
Days
35Hours
15Minutes
14Seconds
52New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.