Learn More
Invitrogen™ iBind™ Flex Cards
Card for use with iBind™ Flex Western Blot Device.
Brand: Invitrogen™ SLF2010X4
This item is not returnable.
View return policy
Description
- Glass fiber backed card with filer paper stack at base
- Quantity: 40 Cards (4 boxes of 10)
Specifications
Store at room temperature. | |
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots | |
Western Blot | |
Nitrocellulose, PVDF | |
∼2.5 hr. | |
Card | |
Room Temperature |
40 Cards (10 x 4) | |
Chromogenic, Chemiluminescence, Fluorescence | |
iBind Flex Western Device | |
iBind Flex | |
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card | |
40 Cards (10 x 4) |
For Research Use Only. Not for use in diagnostic procedures.