missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Solution Kit

Detection kit for use with iBind™ Flex Western System

Brand:  Invitrogen™ SLF2020X4

347.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15901907

  • £369.00 / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

• Optimized solutions with ideal viscosity for blocking, washing, and antibody dilution
• Long shelf life—store iBind Flex Solution Kit for 12 months at 4°C
• Better antigen-antibody interaction—solutions flow between the iBind Flex Card and membrane for maximum interaction between protein and antibody

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flex Solution Kit
iBind Flex Western Device
Qualified for Ambient or Wet Ice
PVDF, Nitrocellulose
4 kits
iBind Flex 5X buffer (400 mL)
iBind Flex 100X additive (12 x 1.7 mL)

Store at 2–8°C.

10 midi blots or 20 mini blots
Colorimetric, Chromogenic, Chemiluminescence
iBind™
Western Blotting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.