Learn More
Invitrogen™ iBind™ Solution Kit
For use with the iBind Western Device
Brand: Invitrogen™ SLF1020
58.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Includes: Kit contains sufficient solution for 10 mini-membrane reactions.
Requires : iBind Western Device
This item is not returnable. View return policy
Description
The automated iBind Western Device performs every step in the Western processing workflow from blocking to washes to antibody incubations. The iBind solutions act as the blocking, washing, and dilution solutions in the iBind immunodetection process and have been optimized for ideal viscosity. Each solution is released from the iBind device wells to the iBind Card through sequential lateral flow (SLF); the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.
- Optimized solutions with ideal viscosity for blocking, washing, and antibody dilution
- Long shelf life—store iBind Solutions Kit for 12 months at 4°C
- Better antigen-antibody interaction—solutions flow between the iBind Card and membrane for maximum interaction between protein and antibody
- Brings automation and simplicity to the Western protocol
Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications
Store at 2–4°C. | |
Western Blot | |
Nitrocellulose, PVDF | |
Solution Kit | |
1 kit |
Colorimetric, Chromogenic, Chemiluminescence | |
iBind Western Device | |
iBind | |
10 mini membrane reactions | |
Protein |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3020855 | Certificate of Analysis | 31/01/2025 | SLF1020,SLF1020_LITL18 PROMO |
2465007 | Certificate of Analysis | 29/01/2025 | SLF1020,SLF1020_LITL18 PROMO |
2483948 | Certificate of Analysis | 29/01/2025 | SLF1020,SLF1020_LITL18 PROMO |
2491552 | Certificate of Analysis | 29/01/2025 | SLF1020,SLF1020_LITL18 PROMO |
2514069 | Certificate of Analysis | 29/01/2025 | SLF1020,SLF1020_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.