missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, mini
Used to transfer proteins with the iBlot 2 Gel Transfer Device
£160.00
Specifications
Format | Transfer Stack |
---|---|
Material | Nitrocellulose |
Pore Size | 0.2 μm |
Content And Storage | Store at room temperature. |
Product Line | iBlot |
Description
The iBlot 2 Transfer Stacks are consumable stacks with integrated nitrocellulose transfer membranes for dry blotting of proteins. Each stack contains a copper coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins.
- Increased shelf life
- Reduced and compact packaging
- Complete compatibility with iBlot 2 Gel Transfer Device
- The nitrocellulose membrane (0.2μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer
- Membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
- Protein binding capacity is 209μg/cm2
Transfer Stack Sizes
- The regular size (13 x 8.3cm) of this stack is used for all Novex midi gels (or other midi gel types), two Novex mini gels (or other mini gel types), and E-PAGE™ 48 or 96 gels
- The mini size (8 x 8cm) is used for all Novex mini gels (or other mini gel types)
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Transfer Stack | |
0.2 μm | |
iBlot |
Nitrocellulose | |
Store at room temperature. |
For Research Use Only. Not for use in diagnostic procedures.