missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, regular size
GREENER_CHOICE

iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins.

£196.00

Specifications

Format Transfer Stack
Material Nitrocellulose
Pore Size 0.2 μm
Content And Storage Store at room temperature.
Dimensions (LxW) 13.5 x 8 cm
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15296526
GREENER_CHOICE
View Documents Promotion Details
Invitrogen™
IB23001
10 Stacks
£196.00
Pack of 10
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
 
Description

Description

iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This regular-size stack (13 x 8.3 cm) can be used with midi-size gels or two mini-size gels or E-PAGE 48 or 96 gels.

Nitrocellulose membrane

The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.

Specifications

Specifications

Transfer Stack
0.2 μm
13.5 x 8 cm
Less waste, responsibly packaged, extended life
Nitrocellulose
Store at room temperature.
iBlot
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X10 iBlot® 2 Transfer Stacks, nitrocellulose

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethylene glycol

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.