missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ 3 Transfer Stacks, midi, nitrocellulose

iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments.

Brand:  Invitrogen™ IB33001X3

Additional Details : Weight : 3.90000kg

 View more versions of this product

Product Code. 18785395

  • £484.00 / Pack of 30
Estimated Shipment: 25-07-2024
to see stock.

Description

Description

iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments. Each iBlot 3 Transfer Stack contains copper-fiber electrode sheets, optimized cathode and anode solid buffer gels, and a preconditioned nitrocellulose transfer membrane. This midi-size stack (13 cm x 8.3 cm) can be used with midi-size gels or two mini-size gels.

The iBlot 3 Western Blot Transfer System (combined iBlot 3 Transfer Stacks and iBlot 3 Transfer Device) have been optimized to provide protein transfers in 3–8 minutes that produce higher quality and more reliable western blot result compared to many other transfer methods.

Nitrocellulose membrane
The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. Proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.

Specifications

Specifications

13.5 cm
3 x 10 Stacks
Nitrocellulose
13.5 x 8 cm (Midi)
30 Blots
8 cm
Transfer Stack
0.2 μm
iBlot
Sustainable disposal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.