Learn More
Invitrogen™ iBlot™ 3 Transfer Stacks, mini, nitrocellulose
iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments.
Brand: Invitrogen™ IB33002X3
Additional Details : Weight : 0.50000kg
Description
iBlot 3 Transfer Stacks are used with iBlot 3 Western Blot Transfer Device for the transfer of proteins from an SDS-PAGE gel to a membrane support when performing western blot experiments. Each iBlot 3 Transfer Stack contains copper-fiber electrode sheets, optimized cathode and anode solid buffer gels, and a preconditioned nitrocellulose transfer membrane. This mini-size stack (8 cm x 8 cm) can be used with one mini-size gel.
The iBlot 3 Western Blot Transfer System (combined iBlot 3 Transfer Stacks and iBlot 3 Transfer Device) have been optimized to provide protein transfers in 3–8 minutes that produce higher quality and more reliable western blot result compared to many other transfer methods.
Nitrocellulose membrane
The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. Proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.
Specifications
8 cm | |
3 x 10 Stacks | |
Nitrocellulose | |
8 x 8 cm (Mini) | |
30 Blots |
8 cm | |
Transfer Stack | |
0.2 μm | |
iBlot | |
Sustainable disposal |
For Research Use Only. Not for use in diagnostic procedures.