Learn More
Ibrutinib-biotin, MedChemExpress
MedChemExpress Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.
Brand:  Medchem Express HY-100342
Encompass_Preferred
 View more versions of this product
Product Code. 16445697
  • £901.45 / 5mg
Explore more special offers
Description
Description
Ibrutinib-biotin is a probe that consists of Ibrutinib linked to biotin via a long chain linker, extracted from patent WO2014059368A1 Compound 1-5, has an IC50 of 0.755-1.02 nM for BTK.
Specifications
Specifications
Ibrutinib-biotin
Colorless
C56H80N12O9S
DMSO : 50 mg/mL (45.56 mM; Need ultrasonic)
1097.37
98.04%
Oil
1599432-18-4
Research
5 mg
O=C(CCCC(NCCCOCCOCCOCCCNC(CCCC[C@@H]1SC[C@H](N2)[C@@H]1NC2=O)=O)=O)NCCCCN(C)C/C=C/C(N3CCC[C@@H](N4N=C(C5=CC=C(OC6=CC=CC=C6)C=C5)C7=C(N)N=CN=C74)C3)=O
1097.37
Research
Cancer-Kinase/protease
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibrutinib-biotin, MedChemExpress