missing translation for 'onlineSavingsMsg'
Learn More
Ibuprofen, 99%, Thermo Scientific Chemicals Product Code.: 15853855
compliance-icons

Ibuprofen, 99%, Thermo Scientific Chemicals

Product Code. 15853855
1 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15853855

Brand: Thermo Scientific Acros 258610010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ibuprofen, 2-4-isobutylphenyl propanoic acid, CAS # 15687-27-1, is a laboratory form of the over-the-counter nonsteroidal anti-inflammatory (NSAID) drug.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Ibuprofen (2-4-isobutylphenyl propanoic acid) is a common NSAID that reduces pyrogen release and inflammation
  • Ibuprofen is a racemic mixture that consists of an R-enantiomer and S-enantiomer; the S-enantiomer is proposed to be the stronger acting agent
  • It is a propionic acid derivative, a short-chain saturated fatty acid bound to ethane
  • Ibuprofen inhibits cyclooxygenases and reduces prostaglandin formation, inhibiting fever and pain

Applications

  • Ibuprofen reduces both local and systemic inflammation; suitable for use in vitro or in vivo to study specific or broad mechanisms
  • Intended for sensitive biochemical applications such as HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

Melting Point 75°C to 78°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2C6H4CH(CH3)COOH
Quantity 1 g
Synonym α-Methyl-4-(2-methylpropyl)benzeneacetic acid, p-Isobutylhydratropic acid, 2-(4-Isobutylphenyl)-propionic acid
Formula Weight 206.27
Percent Purity 98.5% min.
Physical Form Crystalline Powder
Chemical Name or Material Ibuprofen
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0396819 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number258610500,258610100,258610010
Lot NumberA0406854 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number258610500,258610100,258610010
Lot NumberA0414576 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog Number258610500,258610100,258610010
compliance-icons
Product Identifier
  • Ibuprofen
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ibuprofen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ibuprofen, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.