Learn More
Scotsman™ Ice flake maker AF124-Install self contained
Brand:  Scotsman™ AF124+AF124-INSTALL
Product Code. 12813226
  • £6995.00 / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ice flakes are extruded at a temperature just below zero degrees and contain 20% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Produces granular shape ice flakes, 80% hardness
  • Up to 120kg (264,5 lbs) daily production
  • Up to 40kg (88 lbs) ice storage capacity
  • Ice scoop holder safely positioned in ice-storage bin
  • Resistant stainless steel exterior
  • Advanced diagnostics computerised control
  • Front panel in & out airflow (air-cooled model only) for built-in installation
  • Front access condenser air filter, removable and washable (air-cooled version only)
  • Easily accessible, front panel mount, ON-OFF Switch
  • Easy, front panel access to technical / service area of the machine
  • 'Routine Maintenance' visible alarm light on front panel
  • Ergonomically designed ice storage access, with disappearing door
  • Door-closing movement dampening system
  • Vapour-based microbial control system pouch in ice storage area
  • Optional PWD system for remote water drain (easy-fit installation)
  • AgION antimicrobial agent embedded in moulded plastics parts
  • CFC-free R-404A refrigerant
  • Two years manufacturer warranty on defective components
Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Scotsman™ Ice flake maker AF124-Install self contained > includes installation, electronic controls, stainless steel finish 230V 50Hz 950mm x 605mm x 915mm (w x d x h) 120kg/24hrs max production, 40kg bin capacity Scotsman