missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICI 199,441 hydrochloride, Tocris Bioscience™
Potent κ agonist
£110.15 - £385.12
Chemical Identifiers
CAS | 115199-84-3 |
---|---|
Molecular Formula | C21H25Cl3N2O |
Molecular Weight (g/mol) | 427.794 |
InChI Key | VFLWVWZSDBTGQJ-VEIFNGETSA-N |
Synonym | ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s |
PubChem CID | 3082717 |
IUPAC Name | 2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride |
SMILES | CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30001129
|
Tocris Bioscience
0778/10 |
10 mg |
£110.15
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18085652
|
Tocris Bioscience
0778/50 |
50 mg |
£385.12
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly potent κ agonist, 146-fold more active than U-50488 (Cat. No. 0495) in vitro.Chemical Identifiers
115199-84-3 | |
427.794 | |
ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s | |
2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride |
C21H25Cl3N2O | |
VFLWVWZSDBTGQJ-VEIFNGETSA-N | |
3082717 | |
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl |
Specifications
115199-84-3 | |
ici-199,441 hydrochloride, ici 199,441 hydrochloride, n-2-n-methyl-3,4-dichlorophenylacetamido-2-phenylethyl pyrrolidine, s-3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl benzeneacetamide monohydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-1-pyrrolidinyl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-yl ethyl acetamide hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1s-1-phenyl-2-pyrrolidin-1-ylethyl acetamide hydrochloride, benzeneacetamide, 3,4-dichloro-n-methyl-n-1-phenyl-2-1-pyrrolidinyl ethyl-, monohydrochloride, s | |
CN(C(CN1CCCC1)C2=CC=CC=C2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl | |
427.794 | |
427.8 | |
ICI 199, 441 hydrochloride |
C21H25Cl3N2O | |
VFLWVWZSDBTGQJ-VEIFNGETSA-N | |
2-(3,4-dichlorophenyl)-N-methyl-N-[(1S)-1-phenyl-2-pyrrolidin-1-ylethyl]acetamide;hydrochloride | |
3082717 | |
>99% |
Safety and Handling
Recommended Storage : Store at Room Temperature