missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Goat anti-Rabbit, CoraLite594, Secondary Antibody, Proteintech
Goat anti-Rabbit Affinipure Secondary Antibody
Brand: Proteintech SA00013-4-500UL
This item is not returnable.
View return policy
Specifications
IgG(H+L) | |
CoraLite 594 | |
Goat | |
500 μL | |
Rabbit | |
Liquid |
Immunofluorescence, Flow Cytometry | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |