missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Goat anti-Rabbit, CoraLite594, Secondary Antibody, Proteintech

Goat anti-Rabbit Affinipure Secondary Antibody

£40.00 - £84.00

Specifications

Antigen IgG(H+L)
Applications Immunofluorescence, Flow Cytometry
Conjugate CoraLite 594
Form Liquid
Host Species Goat
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17670397
View Documents
Proteintech
SA00013-4-200UL
200 μL
£40.00
200µL
Please to purchase this item. Need a web account? Register with us today!
17680397
View Documents
Proteintech
SA00013-4-500UL
500 μL
£84.00
500µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

IgG(H+L)
CoraLite 594
Goat
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Immunofluorescence, Flow Cytometry
Liquid
Rabbit
Secondary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications