missing translation for 'onlineSavingsMsg'
Learn More

Cytiva IgG Sepharose™ 6 Fast Flow Medium

Use for rapid purification of protein A and protein A fusion conjugates. IgG Sepharose™ 6 Fast Flow Medium achieves the highest flowrates.

Brand:  Cytiva 17-0969-01

Code : GE

Additional Details : Weight : 0.05900kg


Product Code. 11574955

  • £504.00 / 10mL
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Designed for the rapid purification of protein A and protein A fusion conjugates
  • IgG coupled to the highly cross-linked 6% agarose matrix Sepharose 6 Fast Flow
  • Binds at least 2mg protein A/mL medium
  • Suited for tandem affinity purification (TAP) of protein complexes

Order Info

Supplied in 10mL containers

Specifications

Specifications

10mL
90 μm
Sepharose FF Media
5cm
5.9 in.
Human IgG
3 to 10
Protein A and Protein G Conjugates
400 cm/hr.
AKTA™ systems
Solid Bead
Rapid purification of protein A and protein A fusion conjugates, tandem affinity purification (TAP) of protein complexes
15cm
14.5psi
6% Highly Cross-linked Agarose
2° to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • IgG Sepharose 6 Fast Flow, 10 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers