Learn More
Invitrogen™ IL-2 Rat ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-M205
This item is not returnable.
View return policy
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
P17108 | |
116562 | |
2 x 96 Tests | |
Refer to product documentation for component specific storage temperature. |
ELISA | |
IL2 | |
RUO | |
Rat |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
Invitrogen™ IL-2 Rat ELISA Development Kit (ABTS), PeproTech®, PeproTech® > 2 x 96 Tests
Spot an opportunity for improvement?Share a Content Correction